Learn More
o-Phenylenediamine, 98%
Description
o-phenylenediamine is used to prepare tiabendazole, pyrazinamide, morinamide, clemizole and chlormidazole. It is involved in the synthesis of benzimidazole by reacting with carboxylic acids and their derivatives. It is also used to prepare quinoxalinedione and mercaptoimidazoles by condensation with dimethyl oxalate and xanthate esters respectively. It reacts with nitrous acid to get benzotriazole, which finds application as a corrosion inhibitor. It is actively involved in the preparation of pharmaceuticals by reacting substituted o-phenylenediamine with various diketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
o-phenylenediamine is used to prepare tiabendazole, pyrazinamide, morinamide, clemizole and chlormidazole. It is involved in the synthesis of benzimidazole by reacting with carboxylic acids and their derivatives. It is also used to prepare quinoxalinedione and mercaptoimidazoles by condensation with dimethyl oxalate and xanthate esters respectively. It reacts with nitrous acid to get benzotriazole, which finds application as a corrosion inhibitor. It is actively involved in the preparation of pharmaceuticals by reacting substituted o-phenylenediamine with various diketones.
Solubility
Soluble in alcohol, chloroform, ether, slightly soluble in water. Soluble in hot water.
Notes
Moisture and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 100°C to 103°C |
| Density | 1.27 |
| Boiling Point | 256°C to 258°C |
| Flash Point | 136°C (277°F) |
| Assay Percent Range | 98% |
| Quantity | 5000 g |
| UN Number | UN1673 |
| Beilstein | 606074 |
| Sensitivity | Air Sensitive |
| Merck Index | 14,7284 |
| Show More |
Safety and Handling
| Hazard Category | H301-H312+H332-H317-H319-H341-H351-H500 |
| Hazard Statement | GHS H Statement H301-H341-H351-H312-H332-H319-H317 Toxic if swallowed. Suspected of causing genetic defects. Suspected of causing cancer. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLENEDIAMINES |
| EINECSNumber | 202-430-6 |
| RTECSNumber | SS7875000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.