Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich o-Phenylenediamine

Catalog No. 501839451
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501839451 5 g
501839449 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501839451 Supplier Sigma Aldrich Supplier No. P239385G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

o-Phenylenediamine
TRUSTED_SUSTAINABILITY

Specifications

CAS 95-54-5
Melting Point 100°C to 102°C
Boiling Point 256°C to 258°C
Molecular Formula C6H8N2
Linear Formula C6H8N2
MDL Number MFCD00007721
Quantity 5 g
Synonym 1,2-Diaminobenzene; 1,2-Phenylenediamine; OPD
Molecular Weight (g/mol) 108.14
Percent Purity 99.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.