Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Phosphoric Acid, 85% (Certified ACS), Fisher Chemical™

Catalog No. A2424 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Flint Glass Bottle
Glass Bottle
Plastic Bottle
Safe-Cote™ Glass
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7664-38-2, 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

pH 1
Melting Point 21°C
Boiling Point 158°C
Density 1.680g/cm³
Heavy Metals (as Pb) 0.001% max.
Identification Pass Test
Packaging Glass Bottle
Reducing Substances Pass Test
Vapor Pressure 2 hPa at 20°C
Normality about 44.6
Assay Percent Range ≥85.0 w/w %
Quantity 4 L
Physical Form Liquid
UN Number 1805
Formula Weight 98
Percent Purity ≥85.0% (w/w)
Grade Certified ACS
Chemical Name or Material o-Phosphoric Acid, 85%
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:3
Flammability:0
Instability:0
DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.