Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ o-Phosphoric Acid Solution, 85%
SCI_ED

Catalog No. S25470A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
ACS Grade
Reagent Grade
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25470A 1 L ACS Grade
S25470 100 mL ACS Grade
S25470B 2.5 L Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25470A

High-purity 85% o-Phosphoric Acid solution for educational laboratory experiments and demonstrations.

  • Assay: Greater than 85% purity
  • Grade: ACS, reagent grade suitable for education
  • Physical Form: Colorless liquid
  • Density: 1.68 g/mL
  • Quantity: 2.5 liters
  • Molecular Formula: H3PO4
  • Formula Weight: 98 g/mol
  • Packaging: Secure reagent-grade container
  • Use: Designed for a wide range of classroom and instructional laboratory applications
  • Note: Educational use only – no Certificate of Analysis (C of A) available

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5 , 7664-38-2 , 7732-18-5 , 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Color Colorless
Density 1.68
Packaging ACS Grade
Assay Percent Range >85%
Quantity 1 L
Linear Formula H3PO4
Formula Weight 98
Grade ACS
Chemical Name or Material o-Phosphoric Acid Solution, 85%
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 8, III, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.