Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Phosphorylethanolamine 98.0+%, TCI America™
SDP

Catalog No. P0279100G
Click to view available options
Quantity:
25 g
100 g

Chemical Identifiers

CAS 1071-23-4
Molecular Formula C2H8NO4P
Molecular Weight (g/mol) 141.063
MDL Number MFCD00008178
InChI Key SUHOOTKUPISOBE-UHFFFAOYSA-N
Synonym o-phosphoethanolamine, o-phosphorylethanolamine, phosphoethanolamine, o-phosphocolamine, colamine phosphate, phosphorylethanolamine, ethanolamine phosphate, phosphonoethanolamine, ethanolamine o-phosphate, mono 2-aminoethyl phosphate
PubChem CID 1015
ChEBI CHEBI:17553
IUPAC Name 2-aminoethyl dihydrogen phosphate
SMILES C(COP(=O)(O)O)N

Specifications

Melting Point 235°C
Color White-Yellow
Quantity 100 g
UN Number 1759
Formula Weight 141.06
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material O-Phosphorylethanolamine
EINECSNumber (2)-2917
RTECSNumber KJ5830000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.