Learn More
Ritmed™ O.R. Towels Prewashed Towels, Non-Sterile
Catalog No. 21300045
Click to view available options
Color:
Blue
Green
White
Includes:
100 Towels
400 Towels
100 % Cotton muslin operation room towels with 17 x 25 in. used to absorb biological fluids
  • Ritmed™ O.R. Towels are intended for general use in an operating room or other medical environments
  • The towels are prewashed, 100% cotton with latex free feature which makes them ready for immediate use and free of manufacturing residue
  • They are labeled as non-sterile, meaning they are not suitable for use in procedures requiring a sterile field
  • This product is ideal for non-invasive applications or for use where sterilization is performed at the point of use
  • The non-sterile nature makes them a cost-effective option for a variety of tasks where sterility is not the primary requirement
  • Ritmed™ offers a reliable and pre-processed towel for general medical and surgical use
Specifications
Product Type Towel
Color White
Material Cotton
Sterility Non-sterile
Autoclavable Not Autoclavable
Includes 400 Towels
Dimensions (L x W) 17 x 25 in.
Disposable Single-use
Latex Free Yes
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.