Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthware™ O-ring Kits for High Vacuum Valve Plugs

Shop All Kemtech Products


Click to view available options
For Use With (Equipment):
S540004
S540008
S540012
Includes:
R007(x1) and R010(x2)
R108(x1) and R110(x2)
R110(x1) and R112(x2)
Specifications
Specifications
| Color | Brown |
| For Use With (Equipment) | S540004 |
| Material | Viton |
| Includes | R007(x1) and R010(x2) |
| Quantity | 3 Pk. |
| Type | O-Ring Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction