Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O,S-Diethyl methylphosphonothioate, 97%, Thermo Scientific Chemicals

Catalog No. AA4446504
Click to view available options
Quantity:
0.5 g
2 g

O,S-Diethyl methylphosphonothioate is used as an inhibitor of cholinesterase. It is also used in pharmaceuticals, intermediates and fine chemicals. It also serves as phosphorylating and phosphitylating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2511-10-6
MDL Number MFCD01705977

Specifications

Density 1.094 g/mL
Boiling Point 95°C to 97°C (10 mmHg)
Molecular Formula C14H12N2OS
Refractive Index 1.474
Quantity 2 g
UN Number UN3278
Synonym methylene violet, dimethylthionoline, 7-dimethylamino phenothiazin-3-one, 3h-phenothiazin-3-one, 7-dimethylamino, unii-y3g6pd8nrb, y3g6pd8nrb, methylene violet biological stain, 7-dimethylamino-3h-phenothiazin-3-one, methylenviolett, methylene violet bernthsen
Solubility Information Immiscible with water.
InChI Key ALJHHTHBYJROOG-UHFFFAOYSA-N
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=O)C=C3S2
IUPAC Name 7-(dimethylamino)phenothiazin-3-one
Molecular Weight (g/mol) 256.32
PubChem CID 73024
Formula Weight 168.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material O,S-Diethyl methylphosphonothioate
Show More Show Less
Hazard Category H300+H310+H330
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
RTECSNumber TB1085000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.