Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-tert-Butyl-N-Fmoc-L-beta-homoserine, 95%

Catalog No. AAH5206503
Click to view available options
Quantity:
250 mg
1 g

CAS: 203854-51-7 | C23H27NO5 | 397.471 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 203854-51-7
Molecular Formula C23H27NO5
Molecular Weight (g/mol) 397.471
MDL Number MFCD01862865
InChI Key SKYFRYZPXHNPOK-OAHLLOKOSA-N
Synonym fmoc-beta-hoser tbu-oh, fmoc-o-t-butyl-l-beta-homoserine, fmoc-l-beta-homoserine otbu, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy butanoic acid, 3r-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, ambotzfaa6740, fmoc-hoser tbu-oh, fmoc-??-hoser tbu-oh, fmoc-beta-homoser tbu-oh, fmoc-i 2-hoser tbu-oh
PubChem CID 2761544
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]butanoic acid
SMILES CC(C)(C)OCC(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 96°C to 98°C
Quantity 1 g
Beilstein 8014825
Formula Weight 397.47
Percent Purity 95%
Chemical Name or Material O-tert-Butyl-N-Fmoc-L-beta-homoserine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.