Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-tert-Butyl-N-Fmoc-L-beta-homotyrosine, 95%

Catalog No. AAH5219203
Click to view available options
Quantity:
250 mg
1 g

CAS: 219967-69-8 | C29H31NO5 | 473.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 219967-69-8
Molecular Formula C29H31NO5
Molecular Weight (g/mol) 473.57
MDL Number MFCD01862862
InChI Key FKUBPXDMQFEPGP-CYLJNIGPNA-N
Synonym fmoc-l-beta-homotyrosine otbu, fmoc-o-t-butyl-l-beta-homotyrosine, fmoc-o-tert-butyl-l-beta-homotyrosine, fmoc-beta-homotyr tbu-oh, 3s-4-4-tert-butoxy phenyl-3-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, ambotzfaa6770, fmoc-homotyr tbu-oh, fmoc-beta-homotyr otbu-oh, tmba037, 3s-3-9h-fluoren-9-ylmethoxycarbonylamino-4-4-2-methylpropan-2-yl oxy phenyl butanoic acid
PubChem CID 2761556
SMILES CC(C)(C)OC1=CC=C(CC[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1

Specifications

Quantity 1 g
Beilstein 8236890
IUPAC Name (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[4-[(2-methylpropan-2-yl)oxy]phenyl]butanoic acid
Formula Weight 473.57
Percent Purity 95%
Chemical Name or Material O-tert-Butyl-N-Fmoc-L-beta-homotyrosine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.