Learn More
o-Tolidine dihydrochloride, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 130.0°C |
| Color | Beige to Beige-Gray |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (on dry substance) (Argentometry) |
| Packaging | Plastic bottle |
| Quantity | 2.5 kg |
| Merck Index | 15,9675 |
| Formula Weight | 285.2 |
| Percent Purity | 99% |
| Physical Form | Crystalline Powder |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause cancer. Harmful if swallowed. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. |
| EINECSNumber | 210-322-5 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.