Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolidine, Powder, Reagent, Spectrum™ Chemical
SDP

Catalog No. 18604913 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle

Catalog No. 18-604-913

Supplier: Spectrum Chemical Manufacturing Corporation T1084100GM

Only null left
Add to Cart
Add to Cart

T1084, 119-93-7, C14H16N2

Spectrum™ Chemical o-Tolidine, Powder, Reagent is a commercially important aromatic amine used to make dyes. It is also used in the production of certain elastomers and as a reagent and indicator in analytical forensic and clinical chemistry. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 119-93-7
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.30
InChI Key NUIURNJTPRWVAP-UHFFFAOYSA-N
IUPAC Name 3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diamine
SMILES CC1=CC(=CC=C1N)C1=CC=C(N)C(C)=C1

Specifications

CAS Max % 1
Melting Range 125-132°C
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 97.5%
Grade Reagent
Physical Form Crystalline Powder or Lumps
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.