Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolidine Reagent, 0.135% (w/v) in 15% (v/v) Hydrochloric Acid, Ricca Chemical
SDP

Catalog No. 864032 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L

Specifications

Chemical Name or Material o-Tolidine Reagent
Color Colorless to Green-Yellow
CAS 7732-18-5, 7647-01-0, 612-82-8
CAS Min % 83.0, 14.72, 0.11
CAS Max % 87.0, 15.32, 0.12
Synonym dihydrogen oxide, dihydrogen monoxide
Percent Purity 0.135%
Grade Laboratory
Packaging Amber Glass Bottle
Methods APHA for Chlorine
Solvent or Matrix 15% (v/v) Hydrochloric Acid
pH <1
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.135% (w/v)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.