Learn More
o-Tolualdehyde, 98%, stab. with 0.1% hydroquinone
Description
o-tolualdehyde was used in determination of alkenal-2,4-dinitrophenylhydrazones by HPLC by addition of phosphoric acid. It produces to give a single diastereomer of the tetrahydronaphthalene derivative under UV irradiation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
o-tolualdehyde was used in determination of alkenal-2,4-dinitrophenylhydrazones by HPLC by addition of phosphoric acid. It produces to give a single diastereomer of the tetrahydronaphthalene derivative under UV irradiation.
Solubility
Soluble in carbon tetrachloride, ethanol, ethyl ether, benzene. Not miscible in water.
Notes
Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.

Specifications
Specifications
Melting Point | -35°C |
Boiling Point | 199°C to 200°C |
Flash Point | 67°C (152°F) |
Odor | Almond-like |
Quantity | 500 g |
Beilstein | 605841 |
Merck Index | 14,9527 |
Formula Weight | 120.15 |
Percent Purity | 98% |
Chemical Name or Material | o-Tolualdehyde, Stabilized with 0.1% hydroquinone |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement H301-H227-H315-H319-H335 Toxic if swallowed. Combustible liquid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement P210-P261-P301+P310-P305+P351+P338-P405-P501a Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.