Learn More
o-Toluidine, 99%, Thermo Scientific Chemicals
Description
o-Toluidine is used in the production of dyes, as a presumptive test for blood in forensic science. And it is also used in determination of glucose in biological materials, as a precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
o-Toluidine is used in the production of dyes, as a presumptive test for blood in forensic science. And it is also used in determination of glucose in biological materials, as a precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.
Solubility
Soluble in water(15g/L).
Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | -24°C |
| Density | 1.004 |
| Boiling Point | 199°C to 200°C |
| Flash Point | 85°C (185°F) |
| Odor | Unpleasant |
| Refractive Index | 1.572 |
| Quantity | 1000 g |
| UN Number | UN1708 |
| Beilstein | 741981 |
| Sensitivity | Air and light sensitive |
| Show More |
Safety and Handling
| Hazard Category | H227-H301+H331-H312-H315-H318-H341-H350 |
| Hazard Statement | GHS H Statement H301-H331-H350-H227-H319 Toxic if swallowed. Toxic if inhaled. May cause cancer. Combustible liquid. Causes serious eye irritation. |
| Precautionary Statement | P201-P202-P210-P235-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOLUIDINES, LIQUID |
| EINECSNumber | 202-429-0 |
| RTECSNumber | XU2975000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.