Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolyl Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. T335725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T335725G 25 g
T33575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T335725G Supplier TCI America Supplier No. T335725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66107-34-4
Molecular Formula C8H7F3O3S
Molecular Weight (g/mol) 240.20
MDL Number MFCD29089375
InChI Key RBOGJRXPKONDGC-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid o-Tolyl Ester, o-Tolyl Triflate
PubChem CID 572025
IUPAC Name 2-methylphenyl trifluoromethanesulfonate
SMILES CC1=CC=CC=C1OS(=O)(=O)C(F)(F)F

Specifications

Color Colorless
Quantity 25 g
Formula Weight 240.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material o-Tolyl Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.