Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolylacetonitrile, 98+%

Catalog No. p-7022155
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 22364-68-7 | C9H9N | 131.178 g/mol

o-Tolylacetonitrile is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Tolylacetonitrile is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic and chemical intermediate.

Solubility
Slightly soluble in water. Soluble in benzene, ethyl ether, ethanol.

Notes
Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22364-68-7
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00001904
InChI Key WMGVPDQNPUQRND-UHFFFAOYSA-N
Synonym 2-methylbenzyl cyanide, 2-o-tolyl acetonitrile, o-tolylacetonitrile, 2-methylphenylacetonitrile, o-methylphenylacetonitrile, o-xylyl cyanide, o-methylbenzyl cyanide, acetonitrile, o-tolyl, benzeneacetonitrile, 2-methyl, 2-2-methylphenyl acetonitrile
PubChem CID 31155
ChEBI CHEBI:27982
IUPAC Name 2-(2-methylphenyl)acetonitrile
SMILES CC1=CC=CC=C1CC#N

Specifications

Density 1.01
Boiling Point 211°C to 213°C
Flash Point >110°C (230°F)
Odor Almond-like
Refractive Index 1.5275
Quantity 5 g
UN Number UN3276
Beilstein 907182
Solubility Information Slightly soluble in water. Soluble in benzene,ethyl ether,ethanol.
Formula Weight 131.18
Percent Purity ≥98%
Chemical Name or Material o-Tolylacetonitrile
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 244-937-5
RTECSNumber AM2290000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.