Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Xylene, HPLC Grade, 96% min

Catalog No. p-4408302
Click to view available options
Quantity:
1 L
4 L
4 x 1 L

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.04% | CAS: 95-47-6 | C8H10 | 106.17 g/mol

o-Xylene is largely used in the production of phthalic anhydride, and is generally extracted by distillation from a mixed Xylene stream in a plant primarily designed for p-Xylene production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-47-6
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
PubChem CID 7237
ChEBI CHEBI:28063
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

Melting Point -25°C
Density 0.877
Boiling Point 143°C to 145°C
Quantity 1 L
Flash Point 32°C (89°F)
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.505
Specific Gravity 0.877
Linear Formula (CH3)2C6H4
UN Number UN1307
Beilstein 1815558
Solubility Information Soluble in ethanol,diethyl ether,water(0.2g/L).
Formula Weight 106.17
Percent Purity ≥96%
Grade HPLC
Chemical Name or Material o-Xylene
Show More Show Less
Hazard Category H226-H304-H312+H332-H315-H319-H335-H336-H373
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 202-422-2
RTECSNumber ZE2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.