Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Xylene, 99%

Catalog No. aaa11358ae
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 95-47-6 | C8H10 | 106.17 g/mol

o-Xylene is widely used in the production of phthalic anhydride. It is also used in fuels as well as fuel additives. It can be used in the production of isophthalic acid and terephthalic acid. It is utilized in polyvinyl chloride production, alkyl resin and unsaturated polyester resins. It is also used as a solvent in paint, varnish, glue, printing ink, dyes and in rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Xylene is widely used in the production of phthalic anhydride. It is also used in fuels as well as fuel additives. It can be used in the production of isophthalic acid and terephthalic acid. It is utilized in polyvinyl chloride production, alkyl resin and unsaturated polyester resins. It is also used as a solvent in paint, varnish, glue, printing ink, dyes and in rubber.

Solubility
Miscible with ethanol, diethylether. Immiscible with water.

Notes
Incompatible with oxidizing agents and strong acids
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-47-6
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
PubChem CID 7237
ChEBI CHEBI:28063
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

Melting Point -25°C
Density 0.877
Boiling Point 143°C to 145°C
Quantity 100 mL
Flash Point 32°C (89°F)
Refractive Index 1.505
Linear Formula (CH3)2C6H4
UN Number UN1307
Beilstein 1815558
Merck Index 14,10081
Solubility Information Miscible with ethanol,diethylether. Immiscible with water.
Formula Weight 106.17
Percent Purity 99%
Chemical Name or Material o-Xylene
Show More Show Less
Hazard Category H226-H304-H312+H332-H315-H319-H335-H336-H373
Hazard Statement GHS H Statement
H226-H312-H332-H315
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES
EINECSNumber 202-422-2
RTECSNumber ZE2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.