Learn More
o-Xylene, 99%
Description
o-Xylene is widely used in the production of phthalic anhydride. It is also used in fuels as well as fuel additives. It can be used in the production of isophthalic acid and terephthalic acid. It is utilized in polyvinyl chloride production, alkyl resin and unsaturated polyester resins. It is also used as a solvent in paint, varnish, glue, printing ink, dyes and in rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
o-Xylene is widely used in the production of phthalic anhydride. It is also used in fuels as well as fuel additives. It can be used in the production of isophthalic acid and terephthalic acid. It is utilized in polyvinyl chloride production, alkyl resin and unsaturated polyester resins. It is also used as a solvent in paint, varnish, glue, printing ink, dyes and in rubber.
Solubility
Miscible with ethanol, diethylether. Immiscible with water.
Notes
Incompatible with oxidizing agents and strong acids
Specifications
Specifications
| Melting Point | -25°C |
| Density | 0.877 |
| Boiling Point | 143°C to 145°C |
| Quantity | 2500 mL |
| Flash Point | 32°C (89°F) |
| Refractive Index | 1.505 |
| Linear Formula | (CH3)2C6H4 |
| UN Number | UN1307 |
| Beilstein | 1815558 |
| Merck Index | 14,10081 |
| Show More |
Safety and Handling
| Hazard Category | H226-H304-H312+H332-H315-H319-H335-H336-H373 |
| Hazard Statement | GHS H Statement H226-H312-H332-H315 Flammable liquid and vapor. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES |
| EINECSNumber | 202-422-2 |
| RTECSNumber | ZE2450000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.