Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Xylylenediphosphonic Acid 97.0+%, TCI America™
SDP

Catalog No. X00751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00751G 1 g
X0075200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00751G Supplier TCI America Supplier No. X00751G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42104-58-5
Molecular Formula C8H12O6P2
Molecular Weight (g/mol) 266.126
MDL Number MFCD22200404
InChI Key ZCVUWZJGYKGTPG-UHFFFAOYSA-N
PubChem CID 12471823
IUPAC Name [2-(phosphonomethyl)phenyl]methylphosphonic acid
SMILES C1=CC=C(C(=C1)CP(=O)(O)O)CP(=O)(O)O

Specifications

Melting Point 350°C
Quantity 1 g
UN Number 3261
Formula Weight 266.13
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material o-Xylylenediphosphonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.