Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ O4I2
GREENER_CHOICESDP

Catalog No. 566450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-645-0 50 mg
56-641-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 56-645-0 Supplier Tocris Bioscience™ Supplier No. 5664/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Oct3/4 inducer; induces expression of pluripotent-associated genes

O4I2 is an Oct3/4 inducer; promotes Oct3/4 expression and stabilizes the Oct3/4 protein in HEK293 cells. Induces expression of pluripotent-associated genes Lin28, Sox2 and Nanog, and suppresses Rex1. Induces 15-fold increase in Oct3/4 expression in human primary fibroblasts.

Specifications

Chemical Name or Material Ethyl 2-[(4-chlorophenyl)amino]-4-thiazolecarboxylate
CAS 165682-93-9
Quantity 50 mg
Target Additional Stem Cell Dedifferentiation Products
Molecular Formula C12H11ClN2O2S
Formula Weight 282.75
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.