Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OA1/GPR143 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11791R

Description
Ocular albinism type 1 protein is a conserved integral membrane protein with seven transmembrane domains. It is expressed in the eye and epidermal melanocytes. Defects in GPR143 are the cause of ocular albinism type 1 (OA1); also known as Nettleship-Falls type ocular albinism. OA1 is an X-linked disorder characterized by severe impairment of visual acuity, retinal hypopigmentation and the presence of macromelanosomes.Specifications
OA1/GPR143 | |
Polyclonal | |
Unconjugated | |
GPR143 | |
G protein-coupled receptor 143; GPR143; G-protein coupled receptor 143; homolog of human ocular albinism 1 (Nettleship-Falls); MOA1; NYS6; OA1; ocular albinism 1; Ocular albinism type 1 protein; Ocular albinism type 1 protein homolog; RGD1565799 | |
Rabbit | |
Protein A | |
RUO | |
4935 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P51810 | |
GPR143 | |
KLH conjugated synthetic peptide derived from human OA1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction