Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oakton Conductivity and TDS Standard, Cole-Parmer™

Catalog No. p-7084700
Click to view available options
Conductivity:
12,880 μS

Oakton Conductivity and TDS Standard is traceable to NIST standard reference materials. Solutions help users calibrate conductivity instruments to ensure accuracy.

  • KCl, NaCl and 442 conversions printed on the bottle for ease of use (excluded on 60 mL bottle, but included in insert)
  • Temperature corrections are printed on the bottle, with certified values listed in tables and referenced to 25°C (excluded on 60 mL bottle, but included in insert)
  • Manufactured under ISO 9000 quality guidelines and traceable to NIST standards
  • If unopened, shelf life is 1 year. If opened recommended shelf life is 4 months
  • Conductivity options range from 1 μS/cm to 200,000 μS/cm

Specifications

Chemical Name or Material Conductivity Standard
Packaging Plastic
Conductivity 12,880 μS
Technique Calibration of Conductivity Instrument
Color Clear
pH 4.7 to 7.8
Boiling Point 100°C (212°F)
For Use With (Equipment) Conductivity Meter
Solubility Soluble
Solution Type Standard Solution
Type Conductivity and TDS Standard
Quantity 500 mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.