Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oakton™ Waterproof Big Display pHTestr™ 30

Catalog No. p-2918825
Click to view available options
Type:
Pocket Tester
pH Tester

Includes

Meter Only package includes meter, electrode, four 1.5V batteries, instructions. Meter Kit package includes meter; electrode; two each of pH 4.01, 7.00, 10.00 buffers and rinse solution in single-use pouches, sample jar, batteries, instructions and hard Plastic carrying case

Versatile, compact design works in a field or laboratory setting.

  • Simultaneous pH and temperature display
  • Temperature measurement from 0.0° to 50.0°C
  • Suitable for most ASTM and EPA test procedures
  • Microprocessor-based, with accurate readings to 0.01pH
  • Replaceable double-junction electrode (S65290 or NC9830226) has extra reference gel for longer electrode life
  • Automatic temperature compensation
  • Push-button three-point calibration
  • Auto buffer recognition: selectable buffer sets
  • Hold function; auto-off; error messages
  • Waterproof, dustproof and floatable; certifiable to IP67 standards

Order Info

  • Ordering Information
  • Meter dimensions: 6-1/2 in.H x 1-1/2 in. dia. (16.5 x 3.8cm). Shipping weight (meter only): 4.5 oz. (125g).

Warranty and Services

One-year warranty.

Specifications

Accuracy (Temperature) ±1°C
Accuracy (pH) ±0.01
Includes Meter, electrode, four 1.5V batteries, instructions
For Use With (Application) Hold function, auto-off, error messages, waterproof, dustproof and floatable
Measurement Modes pH
Calibration Automatic
Range (pH) -1 to 15
Resolution (pH) 0.01
Calibration Points 1 to 3
Temperature Range (Metric) 0°C to 50°C
Resolution (Temperature) 0.1°C
Display Type LCD
Backlight Option None
Certifications/Compliance NIST
IP Rating IP66/67
Warranty 1 Year
Accuracy ±0.1 pH
Resolution 0.1pH
Detection Range -1 to 15 pH
Temperature (English) Operating 32.0°F to 122.0°F
Temperature (Metric) Operating 0°C to 50°C
Type pH Tester
Length (Metric) 16.5 cm
Diameter (English) 1.5 in.
Diameter (Metric) 3.8 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.