Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OAS1 Monoclonal Antibody (C5), Invitrogen™

Catalog No. PIMA543643
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543643 20 μL
PIMA543642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543643 Supplier Invitrogen™ Supplier No. MA543643
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

OAS1 is an interferon inducible protein that may play a role in mediating resistance to virus infection, control of cell growth, differentiation, and apoptosis. It binds double-stranded RNA and polymerizes ATP into PPP(A2'P5'A)N oligomers, which activate the latent RNase L that, when activated, cleaves single-stranded RNAs. This protein is associated with different subcellular fractions such as mitochondrial, nuclear, and rough/smooth microsomal fractions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OAS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene OAS1
Gene Accession No. P00973
Gene Alias (2-5')oligo(A) synthase 1; (2-5')oligo(A) synthase 1A; (2-5')oligo(A) synthetase 1; (2-5')oligo(A) synthetase 1A; 2',5'-oligo A synthetase 1; 2',5'-oligoadenylate synthetase 1, 40/46kDa; 25 oligoadenylate synthetase; 2'-5' oligoadenylate synthetase; 2'-5' oligoadenylate synthetase 1; 2'-5' oligoadenylate synthetase 1A; 2'-5' oligoadenylate synthetase 1G; 2-5A synthase 1; 2-5A synthase 1A; 2-5A synthetase 1; 2-5A synthetase 1A; 2'-5'-oligoadenylate synthase 1; 2'-5'-oligoadenylate synthase 1; LOW QUALITY PROTEIN: 2'-5'-oligoadenylate synthase 1; 2'-5'-oligoadenylate synthase 1A; 2'-5'-oligoadenylate synthetase 1; 2-5-oligoadenylate synthetase 1; 2'-5'-oligoadenylate synthetase 1, 40/46kDa; 2'-5'-oligoisoadenylate synthetase 1; E16 (2'-5') oligo A synthetase; E18/E16; IFI 4; IFI4; IFI-4; L3; LOW QUALITY PROTEIN: 2'-5'-oligoadenylate synthase 1; OAS 1; Oas1; Oas1a; Oas1g; OIAS; Oias1; OIASI; p42 OAS; p46/p42 OAS
Gene Symbols OAS1
Host Species Mouse
Immunogen Ser74-Pro165 (Accession P00973), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4938
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.