Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OAS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582113
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582113 Supplier Invitrogen™ Supplier No. PA582113
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VFLSPLTTFQ DQLNRRGEFI QEIRRQLEAC QRERAFSVKF EVQAPRWGNP RALSFVLSSL QLGEGVEFDV LPAFDALGQL TGGYKPNPQI YVKLIEECTD LQKEGEFSTC FTELQRDFLK QRPTKLKSLI RLVKHWYQNC KK Highest antigen sequence indentity to the following orthologs: Mouse - 70%, Rat - 68%.

OAS1 is an interferon inducible protein that may play a role in mediating resistance to virus infection, control of cell growth, differentiation, and apoptosis. It binds double-stranded RNA and polymerizes ATP into PPP(A2'P5'A)N oligomers, which activate the latent RNase L that, when activated, cleaves single-stranded RNAs. This protein is associated with different subcellular fractions such as mitochondrial, nuclear, and rough/smooth microsomal fractions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OAS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OAS1
Gene Accession No. P00973
Gene Alias (2-5')oligo(A) synthase 1; (2-5')oligo(A) synthase 1A; (2-5')oligo(A) synthetase 1; (2-5')oligo(A) synthetase 1A; 2',5'-oligo A synthetase 1; 2',5'-oligoadenylate synthetase 1, 40/46kDa; 25 oligoadenylate synthetase; 2'-5' oligoadenylate synthetase; 2'-5' oligoadenylate synthetase 1; 2'-5' oligoadenylate synthetase 1A; 2'-5' oligoadenylate synthetase 1G; 2-5A synthase 1; 2-5A synthase 1A; 2-5A synthetase 1; 2-5A synthetase 1A; 2'-5'-oligoadenylate synthase 1; 2'-5'-oligoadenylate synthase 1; LOW QUALITY PROTEIN: 2'-5'-oligoadenylate synthase 1; 2'-5'-oligoadenylate synthase 1A; 2'-5'-oligoadenylate synthetase 1; 2-5-oligoadenylate synthetase 1; 2'-5'-oligoadenylate synthetase 1, 40/46kDa; 2'-5'-oligoisoadenylate synthetase 1; E16 (2'-5') oligo A synthetase; E18/E16; IFI 4; IFI4; IFI-4; L3; LOW QUALITY PROTEIN: 2'-5'-oligoadenylate synthase 1; OAS 1; Oas1; Oas1a; Oas1g; OIAS; Oias1; OIASI; p42 OAS; p46/p42 OAS
Gene Symbols OAS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OAS1. Recombinant protein control fragment (Product #RP-102253).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4938
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.