Learn More
OAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 149551AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-172-9863
Only null left
Add to Cart
Description
Description
OAS1 is an interferon inducible protein that may play a role in mediating resistance to virus infection, control of cell growth, differentiation, and apoptosis. It binds double-stranded RNA and polymerizes ATP into PPP(A2′P5′A)N oligomers, which activate the latent RNase L that, when activated, cleaves single-stranded RNAs. This protein is associated with different subcellular fractions such as mitochondrial, nuclear, and rough/smooth microsomal fractions.
Specifications
Specifications
OAS1
Polyclonal
Unconjugated
OAS1
(2 5)oligo(A) synthase 1, 2 5 oligoadenylate synthase 1, 2 5A synthase 1, E18/E16, IFI 4, OAS1, OIAS, OIASI, p46/p42 OAS
Rabbit
Antigen Affinity Chromatography
RUO
192281, 246730, 4938
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P00973, P11928, Q05961
OAS1, Oas1a
OAS1 Fusion Protein Ag6793
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.