Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OAS2 Antibody (OTI6E6) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273137
Description
OAS2 Monoclonal specifically detects OAS2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
OAS2 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry | |
(2-5')oligo(A) synthase 2, (2'-5')oligo(A) synthetase 2, (2-5')oligo(A) synthetase 2, 2'5' oligoadenylate synthetase 2, 2-5A synthase 2, 2-5A synthetase 2, 2'-5'-oligoadenylate synthase 2, 2'-5'-oligoadenylate synthetase 2 (69-71 kD), 2'-5'-oligoadenylate synthetase 2, 69/71kDa, EC 2.7.7, EC 2.7.7.-, MGC78578, p69 OAS p71 OAS, p69OAS p71OAS | |
Mouse | |
78.6 kDa | |
100 μg | |
4939 | |
Human | |
Lyophilized |
Western Blot, Immunohistochemistry | |
OTI6E6 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
OAS2 | |
Human recombinant protein fragment corresponding to amino acids 60-335 of human OAS2 (NP_002526) produced in E.coli. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction