Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OAS2 Monoclonal Antibody (OTI5E1)
GREENER_CHOICE

Catalog No. PIMA526557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526557 Supplier Invitrogen™ Supplier No. MA526557
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OAS2 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF

This gene encodes a member of the 2-5A synthetase family, essential proteins involved in the innate immune response to viral infection. The encoded protein is induced by interferons and uses adenosine triphosphate in 2'-specific nucleotidyl transfer reactions to synthesize 2',5'-oligoadenylates. These molecules activate latent RNase L, which results in viral RNA degradation and the inhibition of viral replication. The three known members of this gene family are located in a cluster on chromosome 12. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OAS2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI5E1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene OAS2
Gene Accession No. P29728
Gene Alias (2-5')oligo(A) synthase 2; (2'-5')oligo(A) synthetase 2; 2',5'-oligoadenylate synthetase-like 11; 2,5-oligoadenylate synthetase-like 11; 2'-5' oligoadenylate synthetase 2; 2'-5' oligoadenylate synthetase-like 11; 2-5A synthase 2; 2'-5'-oligoadenylate synthase 2; 2'-5'-oligoadenylate synthetase 2; 2'-5'-oligoadenylate synthetase 2, 69/71kDa; A synthetase 2; MGC78578; Oas2; Oasl11; p69 OAS / p71 OAS; p69OAS / p71OAS
Gene Symbols OAS2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 60-335 of OAS2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4939
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.