Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OASIS/CREB3L1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300039MFV450
Description
OASIS/CREB3L1 Polyclonal antibody specifically detects OASIS/CREB3L1 in Human samples. It is validated for ELISASpecifications
OASIS/CREB3L1 | |
Polyclonal | |
ELISA | |
BBF-2 homolog, cAMP responsive element binding protein 3-like 1, cAMP-responsive element-binding protein 3-like protein 1, OASIScyclic AMP-responsive element-binding protein 3-like protein 1, old astrocyte specifically induced substance, Old astrocyte specifically-induced substance | |
Produced in rabbits immunized with purified, recombinant Human OASIS/CREB3L1 (Uniprot#: Q96BA8-1; Glu396- Ser519) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
90993 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction