Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OASL Recombinant Rabbit Monoclonal Antibody (HL2399)
GREENER_CHOICE

Catalog No. FEMA550518
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550518 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550518 Supplier Invitrogen™ Supplier No. MA550518
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

OASL Recombinant Monoclonal Antibody for Western Blot, IHC (P)

2-prime,5-prime oligoadenylates (2-5As) bind to and activate RNase L, leading to degradation of RNA and inhibition of protein synthesis. 2-5As are produced by 2-5A synthetases (OASs), a highly-conserved family of interferon-induced enzymes. The predicted 514-amino acid human p59OASL (2-5A synthetases-like) protein shares a highly conserved N-terminal domain with other OASs. The C-terminal portion of p59OASL contains 2 ubiquitin-like domains. p59OASL is expressed in most tissues, with the highest levels in hematopoietic tissues, colon, and stomach.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OASL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2399
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene OASL
Gene Accession No. Q15646
Gene Alias 2',5'-oligoadenylate synthetase-like 9; 2,5-oligoadenylate synthetase-like 9; 2'-5' oligoadenylate synthetase-like 1; 2'-5'-OAS-related protein; 2'-5'-OAS-RP; 2'-5'-oligoadenylate synthase-like protein; 2'-5'-oligoadenylate synthase-like protein 1; 2'-5'-oligoadenylate synthetase like; 2'-5'-oligoadenylate synthetase-like; 59 kDa 2'-5'-oligoadenylate synthase-like protein; 59 kDa 2'-5'-oligoadenylate synthetase-like protein; 7530414C13Rik; OASL; Oasl1; oasl9; OASLd; p59 OASL; p59OASL; p59-OASL; Thyroid receptor-interacting protein 14; TR-interacting protein 14; TRIP14; TRIP-14
Gene Symbols OASL
Host Species Rabbit
Immunogen Recombinant fragment of human OASL.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8638
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.