Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OAT Antibody (23A2), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP149312AF405
Description
OAT Monoclonal specifically detects OAT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
OAT | |
Monoclonal | |
Alexa Fluor 405 | |
NP_000265 | |
OAT | |
Recombinant human OAT (33-439aa) purified from E. coli. | |
0.1 mL | |
metabolism, Proteases & Other Enzymes | |
4942 | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
23A2 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
GACR, HOGA, OAT ornithine aminotransferase, OATASE, OKT | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human OAT (33-439aa). | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction