Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OAT Antibody (23A2), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP149312AF750
Description
OAT Monoclonal specifically detects OAT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
OAT | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
OAT | |
Recombinant human OAT (33-439aa) purified from E. coli. | |
metabolism, Proteases & Other Enzymes | |
4942 | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
23A2 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
GACR, HOGA, OAT ornithine aminotransferase, OATASE, OKT | |
Mouse | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction