Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OAT Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA527968

Catalog No. PIPA527968


Only null left
Add to Cart

Description

Description

Recommended positive controls: K562, THP-1, Mouse brain, OAT-transfected 293T. Predicted reactivity: Mouse (93%), Rat (93%), Zebrafish (81%), Xenopus laevis (84%), Pig (94%), Chicken (86%), Rhesus Monkey (97%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

OAT is the mitochondrial enzyme ornithine aminotransferase, which is a key enzyme in the pathway that converts arginine and ornithine into the major excitatory and inhibitory neurotransmitters glutamate and GABA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OAT
Polyclonal
Unconjugated
OAT
AI194874; GACR; gyrate atrophy; HOGA; OAT; OATASE; OKT; ornithine aminotransferase; ornithine aminotransferase (gyrate atrophy); Ornithine aminotransferase, hepatic form; Ornithine aminotransferase, mitochondrial; Ornithine aminotransferase, renal form; Ornithine delta-aminotransferase; ornithine-delta-aminotransferase; ornithine--oxo-acid aminotransferase; ornithine-oxo-acid aminotransferase; rOAT; testicular tissue protein Li 130
Rabbit
Antigen affinity chromatography
RUO
18242, 4942
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.28 mg/mL
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
P04181, P29758
OAT
Recombinant fragment corresponding to a region within amino acids 30 and 395 of Human OAT.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.