Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OATP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101905
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101905 Supplier Invitrogen™ Supplier No. PA5101905
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLCO1A2.

The organic anion transporting polypeptides, OATP-A (also designated OATP1, OATP1A2 and SLC21A3) and OATP-C (also designated OATP2, SLC21A6 and LST1), mediate hepatic uptake of cardiac glycosides. The expression of OATP-C, but not OATP-A, is inducible by phenobarbital and pregnenolone-16a-carbonitrile, resulting in the increased capacity of the liver to extract cardiac glycosides from the plasma. OATP-A is expressed in liver and kidney and helps mediate sodium-independent uptake of the anionic steroid conjugates dehydroepiandrosterone sulfate, estradiol-17-glucuronide and prostaglandin. OATP-C is exclusively expressed in liver and is localized to the basolateral hepatocyte membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OATP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLCO1A2
Gene Accession No. P46721
Gene Alias A530084B21; Brain digoxin carrier protein; brain-specific organic anion transporter; membrane transporter; OATP; Oatp1; OATP-1; Oatp1a1; OATP1A2; Oatp1a4; Oatp2; OATPA; OATP-A; OATP-B1; organic anion transporter 2; organic anion transporting polypeptide 1; organic anion transporting polypeptide 1a2; organic anion transporting polypeptide 1a2 variant 1; organic anion transporting polypeptide A; organic anion-transporting polypeptide 1; Slc21a1; SLC21A3; Slc21a5; Slco1a1; SLCO1A2; Slco1a4; SO1A2; Sodium-independent organic anion transporter; sodium-independent organic anion transporter 1; Sodium-independent organic anion-transporting polypeptide 1; sodium-independent organic anion-transporting polypeptide 2; solute carrier family (organic anion transporter) member 1; solute carrier family (organic anion transporter) member 3; solute carrier family 21 (organic anion transporter), member 1; solute carrier family 21 (organic anion transporter), member 3; solute carrier family 21 (organic anion transporter), member 5; solute carrier family 21 member 1; Solute carrier family 21 member 3; solute carrier family 21 member 5; solute carrier family 21, member 1; solute carrier organic anion transporter family member 1A1; Solute carrier organic anion transporter family member 1A2; solute carrier organic anion transporter family member 1A4; solute carrier organic anion transporter family, member 1a1; solute carrier organic anion transporter family, member 1A2; solute carrier organic anion transporter family, member 1a4
Gene Symbols SLCO1A2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLCO1A2(Accession P46721), corresponding to amino acid residues C252-T302.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6579
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.