Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OATP2 Monoclonal Antibody (ESL)
GREENER_CHOICE

Catalog No. MA3934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA3934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA3934 Supplier Invitrogen™ Supplier No. MA3934
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA3-934 detects OATP from human tissue and cells. This antibody does not detect dog, mouse or rat protein. MA3-934 shows no cross-reactivity to other OATP isoforms. MA3-934 has been successfully used in immunofluorescence, western blotting, and immunoprecipitation procedures. Immunofluorescence staining of OATP2 in human liver samples results in a pattern consistent with plasma membrane staining. The MA3-934 immunogen is a synthetic peptide corresponding to residues E(671) S L N K N K H F V P S A G A D S E T H C(691) of human OATP2.

A liver-specific member of the organic anion transporter family. This protein is a transmembrane receptor that mediates the sodium-independent uptake of numerous endogenous compounds including bilirubin, 17-beta-glucuronosyl estradiol and leukotriene C4. This protein is also involved in the removal of drug compounds such as statins, bromosulfophthalein and rifampin from the blood into the hepatocytes. Polymorphisms in the gene encoding this protein are associated with impaired transporter function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OATP2
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone ESL
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 0.09% sodium azide
Gene SLCO1B1
Gene Accession No. Q9Y6L6
Gene Alias HBLRR; liver-specific organic anion transporter 1; LST1; LST-1; OATP1B1; OATP2; OATP-2; OATPC; OATP-C; SLC01B1; SLC21A6; SLCO1B1; Sodium-independent organic anion-transporting polypeptide 2; solute carrier family 21 (organic anion transporter), member 6; Solute carrier family 21 member 6; Solute carrier organic anion transporter family member 1B1; solute carrier organic anion transporter family, member 1B1
Gene Symbols SLCO1B1
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues E(671) S L N K N K H F V P S A G A D S E T H C(691) of human OATP2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10599
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.