Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OATP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584034 Supplier Invitrogen™ Supplier No. PA584034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVPLSYCNSE CNCDESQWEP VCGNNG Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 77%.

SLCO1B3, also known as OATP8 or OATP1B3, is a member of Organic Anion Transporting Polypeptides (OATPs) which are sodium-independent organic anion transporters mediating the uptake of a wide range of structurally diverse endogenous and exogenous compounds including bile acids, hormone conjugates, peptides, toxins, as well as a multitude of therapeutic drugs. SLCO1B3 is specifically expressed in liver under normal condition, while its expression has also been observed in cancer tissues like colon, prostate and pancreas. The molecular mass of SLCO1B3 is highly dependent on the glycosylation modification (deglycosylated form of 65 kDa, fully-glycosylated form of 120 kDa).
TRUSTED_SUSTAINABILITY

Specifications

Antigen OATP8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLCO1B3
Gene Accession No. Q9NPD5
Gene Alias HBLRR; liver-specific organic anion transporter 2; liver-specific organic anion transporter 3TM13; LST2; LST-2; LST3; LST-3TM13; OATP1B3; OATP8; OATP-8; Organic anion transporter 8; organic anion transporter LST-3c; organic anion-transporting polypeptide 8; SLC21A8; SLCO1B3; solute carrier family 21 (organic anion transporter), member 8; Solute carrier family 21 member 8; Solute carrier organic anion transporter family member 1B3; solute carrier organic anion transporter family, member 1B3
Gene Symbols SLCO1B3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLCO1B3. Recombinant protein control fragment (Product #RP-104462).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28234
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.