Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OAZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143282
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143282 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5143282 Supplier Invitrogen™ Supplier No. PA5143282
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

OAZ1 catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. This protein play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. The protein expression is auto-regulated by polyamine-enhanced translational frameshifting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OAZ1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene OAZ1
Gene Accession No. P54368
Gene Alias Antizyme; antizyme 1; AZ1; AZ-1; AZI; OAZ; OAZ1; ODCAC; ODC-Az; Ornithine decarboxylase antizyme 1
Gene Symbols OAZ1
Host Species Goat
Immunogen Peptide with sequence C-SRLTDAKRINWRT (aa 120-132).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4946
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.