Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OAZ2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561570
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561570 Supplier Invitrogen™ Supplier No. PA561570
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PHPLSKIPGG RGGGRDPSLS ALIYKDEKLT VTQDLPVNDG KPHIVHFQYE VTEVKVSSWD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 98%.

Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. The ornithine decarboxylase antizymes play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. Antizyme expression is auto-regulated by polyamine-enhanced translational frameshifting. The antizyme encoded by this gene inhibits ornithine decarboxylase but does not accelerate its degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OAZ2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OAZ2
Gene Accession No. O95190
Gene Alias antizyme 2; AZ2; AZ-2; OAZ2; Oaz2-ps; ODC-Az 2; ornithine decarboxylase antizyme 2; RGD1562933; seizure-related protein 15; Sez15
Gene Symbols OAZ2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OAZ2. Recombinant protein control fragment (Product #RP-98145).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4947
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.