Learn More
Invitrogen™ OAZ2 Polyclonal Antibody

Description
Predicted to react with Mouse and Rat samples.
Specifications
Specifications
| Antigen | OAZ2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | OAZ2 |
| Gene Accession No. | O08608, O95190 |
| Gene Alias | antizyme 2; AZ2; AZ-2; OAZ2; Oaz2-ps; ODC-Az 2; ornithine decarboxylase antizyme 2; RGD1562933; seizure-related protein 15; Sez15 |
| Gene Symbols | OAZ2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.