Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OAZ3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575631
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The antizyme family is a group of small proteins that play a role in cell growth and division by regulating the biosynthesis of polyamines such as putrescine, spermidine and spermine. AZ3, also known as OAZ3 (Ornithine decarboxylase antizyme 3), is a 187 amino acid protein that belongs to the ODC antizyme family. AZ3 probably plays a key role in spermatogenesis by regulating the intracellular concentration of polyamines in haploid germ cells because it binds to, and destabilizes, ornithine decarboxylase. However, mutations in the AZ3 gene are not a common cause of male infertility and the normal AZ3 protein does not accelerate ornithine decarboxylase degeneration. AZ3 expression has a sharp onset in early spermatids, peaks in later stages and is gone in spermatozoa.
Specifications
OAZ3 | |
Polyclonal | |
Unconjugated | |
OAZ3 | |
antizyme 3; AZ3; Az-3; OAZ3; OAZ-t; ODC-Az 3; ornithine decarboxylase antizyme 3; testicular secretory protein Li 31; testis specific ornithine decarboxylase antizyme; TISP15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51686, 53814, 689588 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
A1BPI0, Q9R109, Q9UMX2 | |
OAZ3 | |
Synthetic peptide corresponding to residues in Human OAZ3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction