Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OAZ3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565056
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565056 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565056 Supplier Invitrogen™ Supplier No. PA565056
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VNFQNDRNDRG ALLRAFSYMG FEVVRPDHPA LPPLDNVIFM VYPLERDVGH LPSEPP Highest antigen sequence identity to the following orthologs - mouse 82%, rat 77%.

The antizyme family is a group of small proteins that play a role in cell growth and division by regulating the biosynthesis of polyamines such as putrescine, spermidine and spermine. AZ3, also known as OAZ3 (Ornithine decarboxylase antizyme 3), is a 187 amino acid protein that belongs to the ODC antizyme family. AZ3 probably plays a key role in spermatogenesis by regulating the intracellular concentration of polyamines in haploid germ cells because it binds to, and destabilizes, ornithine decarboxylase. However, mutations in the AZ3 gene are not a common cause of male infertility and the normal AZ3 protein does not accelerate ornithine decarboxylase degeneration. AZ3 expression has a sharp onset in early spermatids, peaks in later stages and is gone in spermatozoa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OAZ3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OAZ3
Gene Accession No. Q9UMX2
Gene Alias antizyme 3; AZ3; Az-3; OAZ3; OAZ-t; ODC-Az 3; ornithine decarboxylase antizyme 3; testicular secretory protein Li 31; testis specific ornithine decarboxylase antizyme; TISP15
Gene Symbols OAZ3
Host Species Rabbit
Immunogen Recombinant Human OAZ3. Recombinant protein control fragment (Product #RP-105822).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51686
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.