Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ OB-1
GREENER_CHOICESDP

Catalog No. 654550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-455-0 50 mg
65-451-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 65-455-0 Supplier Tocris Bioscience™ Supplier No. 6545/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Stomatin-like protein-3 (STOML3) oligomerization inhibitor

OB-1 is a stomatin-like protein-3 (STOML3) oligomerization inhibitor. Reduces STOML3 cluster size in the plasma membrane. Decreases mechanically-induced Piezo1 but not ASIC3 current amplitudes in N2a cells and cultured mouse sensory neurons in vitro. Subcutaneous administration in mice reversibly silences touch receptor activity and inhibits paw withdrawal reflex in a mouse neuropathic pain model.

Specifications

Chemical Name or Material Ethyl 5-(1-methyl-4-nitro-1H-imidazol-5-yloxy)-2-phenylbenzofuran-3-carboxylate
CAS 300803-69-4
Quantity 50 mg
Molecular Formula C21H17N3O6
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.