Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ OB 24 hydrochloride
GREENER_CHOICE

Catalog No. 611910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-191-0 10 mg
61-195-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-191-0 Supplier Tocris Bioscience™ Supplier No. 6119/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective heme oxygenase 1 (HO-1) inhibitor

OB 24 hydrochloride is a potent and selective heme oxygenase 1 (HO-1) inhibitor (IC50 values are 1.9 and >100 μM for HO-1 and HO-2 respectively), reduces protein carbonylation and reactive oxygen species formation in advanced prostate cancer cells (PCA). Inhibits cell proliferation in vitro and PCA tumor growth and lymph node/lung metastases in vivo. Synergizes with Taxol (Cat. No. 1097).

Specifications

Chemical Name or Material 1-[[2-[2-(4-Bromophenyl)ethyl]-1,3-dioxolan-2-yl]methyl]-1H-imidazole hydrochloride
CAS 939825-12-4
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C15H17BrN2O2.HCl
Formula Weight Observed MW: 373.67
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.