Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OBFC1 Antibody (OTI2E4), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273138PEATT594
Description
OBFC1 Monoclonal specifically detects OBFC1 in Human samples. It is validated for Flow Cytometry.Specifications
OBFC1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
STN1 | |
Full length recombinant protein of human OBFC1 (NP_079204) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
OTI2E4 | |
Flow Cytometry | |
AAF44, AAF-44, alpha accessory factor 44, bA541N10.2, CST complex subunit STN1, oligonucleotide/oligosaccharide-binding fold containing 1, Oligonucleotide/oligosaccharide-binding fold-containing protein 1, replication protein A 32 kDa subunit, RP11-541N10.2, RPA-32, STN1FLJ22559, Suppressor of cdc thirteen homolog | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
79991.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction