Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OBFC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581049
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581049 Supplier Invitrogen™ Supplier No. PA581049
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human OBFC1.

OBFC1 is a component of the CST complex, a complex that binds to single-stranded DNA and is required to protect telomeres from DNA degradation. The CST complex binds single-stranded DNA with high affinity in a sequence-independent manner, while isolated subunits bind DNA with low affinity by themselves. In addition to telomere protection, the CST complex has probably a more general role in DNA metabolism at non-telomeric sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OBFC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene STN1
Gene Accession No. Q9H668
Gene Alias 0610009H20Rik; 2310057J23Rik; AAF44; AAF-44; AI413458; alpha accessory factor 44; alpha-accessory factor of 44 kDa; bA541N10.2; CST complex subunit STN1; Obfc1; oligonucleotide/oligosaccharide binding fold containing 1; oligonucleotide/oligosaccharide-binding fold containing 1; oligonucleotide/oligosaccharide-binding fold-containing protein 1; replication protein A 32 kDa subunit; RPA-32; STN1; STN1 subunit of CST complex; STN1, CST complex subunit; suppressor of cdc thirteen homolog
Gene Symbols STN1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human OBFC1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79991
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.