Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OBFC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | OBFC1 |
---|---|
Concentration | 0.08 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111089
|
Invitrogen™
PA5111089 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111090
|
Invitrogen™
PA5111090 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: PVFLAFAKLY IRDILDMKES RQVPGVFLYN GHPIKQVDVL GTVIGVRERD AFYSYGVDDS TGVINCICWK KLNTESVSAA PSAARELSLT SQLKKLQETI EQKTKI.
OBFC1 is a component of the CST complex, a complex that binds to single-stranded DNA and is required to protect telomeres from DNA degradation. The CST complex binds single-stranded DNA with high affinity in a sequence-independent manner, while isolated subunits bind DNA with low affinity by themselves. In addition to telomere protection, the CST complex has probably a more general role in DNA metabolism at non-telomeric sites.Specifications
OBFC1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
STN1 | |
0610009H20Rik; 2310057J23Rik; AAF44; AAF-44; AI413458; alpha accessory factor 44; alpha-accessory factor of 44 kDa; bA541N10.2; CST complex subunit STN1; Obfc1; oligonucleotide/oligosaccharide binding fold containing 1; oligonucleotide/oligosaccharide-binding fold containing 1; oligonucleotide/oligosaccharide-binding fold-containing protein 1; replication protein A 32 kDa subunit; RPA-32; STN1; STN1 subunit of CST complex; STN1, CST complex subunit; suppressor of cdc thirteen homolog | |
STN1 | |
Primary | |
Antigen Affinity Chromatography |
0.08 mg/mL | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H668 | |
79991 | |
Recombinant Protein corresonding to Human OBFC1. Recombinant protein control fragment (Product #RP-97057). | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title