Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OBFC2A Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126639G
Description
OBFC2A Polyclonal antibody specifically detects OBFC2A in Human samples. It is validated for Western BlotSpecifications
OBFC2A | |
Polyclonal | |
Western Blot | |
DKFZp667M1322, FLJ13624, FLJ22833, hSSB2, MGC111163, oligonucleotide/oligosaccharide-binding fold containing 2A, Oligonucleotide/oligosaccharide-binding fold-containing protein 2A, Sensor of single-strand DNA complex subunit B2, Sensor of ssDNA subunit B2, SOSS complex subunit B2, SOSS-B2, SSB2Single-stranded DNA-binding protein 2 | |
The immunogen recognized by this antibody maps to a region between residue 1 to 50 of human oligonucleotide/oligosaccharide-binding fold containing 2A using the numbering given in entry NP_001026886.1 (GeneID 64859). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
64859 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction