Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OBFC2B Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP106561R
Description
OBFC2B Polyclonal antibody specifically detects OBFC2B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
OBFC2B | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
LP3587, MGC2731, oligonucleotide/oligosaccharide-binding fold containing 2B, Oligonucleotide/oligosaccharide-binding fold-containing protein 2B, Sensor of single-strand DNA complex subunit B1, Sensor of ssDNA subunit B1, Single-stranded DNA-binding protein 1, SOSS complex subunit B1, SOSS-B1, SSB1hSSB1 | |
The immunogen recognized by this antibody maps to a region between residue 161 and 211 of human single-stranded DNA-binding protein 1 using the numbering given in entry NP_076973.1 (GeneID 79035). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
79035 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction